accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1092460-75-7

H3CONFFFFFF

CAS RN 1092460-75-7

IUPAC Name: 2-methoxy-4,6-bis(trifluoromethyl)benzonitrile
Synonyms: 2-methoxy-4,6-bis(trifluoromethyl)benzonitrile
Molecular Weight (g/mol): 269.15
Molecular Formula: C10H5F6NO
InChi Key: UEQPQBOSRAOLAK-UHFFFAOYSA-N
SMILES: COC1=CC(=CC(=C1C#N)C(F)(F)F)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results