accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3H3CCH3OOHNOOH

CAS RN 109183-71-3

IUPAC Name: 2-{[(tert-butoxy)carbonyl]amino}-2-cyclohexylacetic acid
Synonyms: [(tert-butoxycarbonyl)amino](cyclohexyl)acetic acid
Molecular Weight (g/mol): 257.33
Molecular Formula: C13H23NO4
InChi Key: QSUXZIPXYDQFCX-UHFFFAOYNA-N
SMILES: CC(C)(C)OC(=O)NC(C1CCCCC1)C(O)=O
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)