CAS RN 1087659-30-0
CAS RN 1087659-30-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6-Chloro-2,3-dimethoxypyridine, TRC
CAS: 1087659-30-0 Molecular Formula: C7H8NO2Cl Molecular Weight (g/mol): 173.6 IUPAC Name: 6-chloro-2,3-dimethoxypyridine SMILES: COc1ccc(Cl)nc1OC