accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

NNSNH2F

CAS RN 1086385-83-2

IUPAC Name: 5-(4-fluorophenyl)-1,2,4-thiadiazol-3-amine
Synonyms: 5-(4-fluorophenyl)-1,2,4-thiadiazol-3-amine
Molecular Weight (g/mol): 195.22
Molecular Formula: C8H6FN3S
InChi Key: MMIMNOLKKPZKTQ-UHFFFAOYSA-N
SMILES: NC1=NSC(=N1)C1=CC=C(F)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results