accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CH3CCH3SiH3CCH3OOH

CAS RN 108534-47-0

IUPAC Name: 4-[(tert-butyldimethylsilyl)oxy]phenol
Synonyms: 4-[(tert-butyldimethylsilyl)oxy]phenol
Molecular Weight (g/mol): 224.38
Molecular Formula: C12H20O2Si
InChi Key: KZTVHIZALLBXMO-UHFFFAOYSA-N
SMILES: CC(C)(C)[Si](C)(C)OC1=CC=C(O)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results