accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

ZnCC

CAS RN 1078-58-6

IUPAC Name: zinc(2+) bis(benzenide)
Synonyms: diphenylzinc
Molecular Weight (g/mol): 219.59
Molecular Formula: C12H10Zn
InChi Key: BUNROVZNGITPIX-UHFFFAOYSA-N
SMILES: [Zn++].C1=CC=[C-]C=C1.C1=CC=[C-]C=C1
Boiling Point 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results