accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1078-28-0

CH3ONCH3

CAS RN 1078-28-0

IUPAC Name: 6-methoxy-2-methylquinoline
Synonyms: 6-methoxy-2-methylquinoline
Molecular Weight (g/mol): 173.22
Molecular Formula: C11H11NO
InChi Key: NAGJQQFMJKMXJQ-UHFFFAOYSA-N
SMILES: COC1=CC=C2N=C(C)C=CC2=C1
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (1)