accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1073353-90-8

BOOH3CCH3H3CH3CHN

CAS RN 1073353-90-8

IUPAC Name: N-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}aniline
Synonyms: N-{[3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]methyl}aniline
Molecular Weight (g/mol): 309.22
Molecular Formula: C19H24BNO2
InChi Key: FMCOCBRSLHQLQR-UHFFFAOYSA-N
SMILES: CC1(C)OB(OC1(C)C)C1=CC(CNC2=CC=CC=C2)=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results