CAS RN 1070-64-0
CAS RN 1070-64-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
6,8-Dichlorooctanoic Acid Ethyl Ester, TRC
CAS: 1070-64-0 Molecular Formula: C10 H18 Cl2 O2 Molecular Weight (g/mol): 241.15 Synonym: Ethyl 6,8-Dichlorooctanoate SMILES: CCOC(=O)CCCCC(Cl)CCCl