accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOFFF

CAS RN 106312-36-1

IUPAC Name: 4-methoxy-2-(trifluoromethyl)benzaldehyde
Synonyms: 4-methoxy-2-(trifluoromethyl)benzaldehyde
Molecular Weight (g/mol): 204.15
Molecular Formula: C9H7F3O2
InChi Key: BVPVUMRIGHMFNV-UHFFFAOYSA-N
SMILES: COC1=CC(=C(C=O)C=C1)C(F)(F)F
Molecular Weight (g/mol) 
  • (2)
  • (4)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results