accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1034248-85-5

CH3OOClF

CAS RN 1034248-85-5

IUPAC Name: 4-ethoxy-2-fluorobenzoyl chloride
Synonyms: 4-ethoxy-2-fluorobenzoyl chloride
Molecular Weight (g/mol): 202.61
Molecular Formula: C9H8ClFO2
InChi Key: FTJONAPGITZXSQ-UHFFFAOYSA-N
SMILES: CCOC1=CC(F)=C(C=C1)C(Cl)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results