accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 10340-23-5

H3COH(Z)

CAS RN 10340-23-5

IUPAC Name: (3Z)-non-3-en-1-ol
Synonyms: (3Z)-non-3-en-1-ol
Molecular Weight (g/mol): 142.24
Molecular Formula: C9H18O
InChi Key: IFTBJDZSLBRRMC-SREVYHEPSA-N
SMILES: CCCCC\C=C/CCO
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (1)
  • (1)