accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

BOOH3CCH3H3CH3CNNH3COOCH3CH3CH3CH3

CAS RN 1032758-82-9

IUPAC Name: tert-butyl N-methyl-N-[3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Synonyms: tert-butyl N-methyl-N-[3-methyl-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridin-2-yl]carbamate
Molecular Weight (g/mol): 348.25
Molecular Formula: C18H29BN2O4
InChi Key: FZESGBZQXNHCBH-UHFFFAOYSA-N
SMILES: CN(C(=O)OC(C)(C)C)C1=C(C)C=C(C=N1)B1OC(C)(C)C(C)(C)O1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results