accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 102422-56-0

FNCO

CAS RN 102422-56-0

IUPAC Name: 1-fluoro-3-(isocyanatomethyl)benzene
Synonyms: 1-fluoro-3-(isocyanatomethyl)benzene
Molecular Weight (g/mol): 151.14
Molecular Formula: C8H6FNO
InChi Key: PHRJTGPFEAUEBC-UHFFFAOYSA-N
SMILES: FC1=CC=CC(CN=C=O)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results