accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3CONH2H2N

CAS RN 102-51-2

IUPAC Name: 4-methoxybenzene-1,2-diamine
Synonyms: 4-methoxybenzene-1,2-diamine
Molecular Weight (g/mol): 138.17
Molecular Formula: C7H10N2O
InChi Key: AGAHETWGCFCMDK-UHFFFAOYSA-N
SMILES: COC1=CC=C(N)C(N)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results