CAS RN 102-48-7
CAS RN 102-48-7
IUPAC Name:
1-(3,4-dimethylphenyl)methanamine
Synonyms:
1-(3,4-dimethylphenyl)methanamine
Molecular Weight (g/mol):
135.21
Molecular Formula:
C9H13N
InChi Key:
PXNRCZQMDSDSHJ-UHFFFAOYSA-N
SMILES:
CC1=CC=C(CN)C=C1C
1
–
1
of
1
results
1
3,4-Dimethylbenzylamine, Thermo Scientific™
CAS: 102-48-7 Molecular Formula: C9H13N Molecular Weight (g/mol): 135.21 MDL Number: MFCD00025577 InChI Key: PXNRCZQMDSDSHJ-UHFFFAOYSA-N PubChem CID: 66888 IUPAC Name: (3,4-dimethylphenyl)methanamine SMILES: CC1=C(C=C(C=C1)CN)C