accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1017779-67-7

CH3OClF

CAS RN 1017779-67-7

IUPAC Name: 1-(4-chloro-3-fluorophenyl)propan-1-one
Synonyms: 1-(4-chloro-3-fluorophenyl)propan-1-one
Molecular Weight (g/mol): 186.61
Molecular Formula: C9H8ClFO
InChi Key: FNPWFFVVAPSNJH-UHFFFAOYSA-N
SMILES: CCC(=O)C1=CC(F)=C(Cl)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results