accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OOOHFFF

CAS RN 1017779-10-0

IUPAC Name: 2-[2-methoxy-6-(trifluoromethyl)phenyl]acetic acid
Synonyms: [2-methoxy-6-(trifluoromethyl)phenyl]acetic acid
Molecular Weight (g/mol): 234.17
Molecular Formula: C10H9F3O3
InChi Key: ZHLWLKKEYBOIHC-UHFFFAOYSA-N
SMILES: COC1=CC=CC(=C1CC(O)=O)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results