accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3OFOH3CF

CAS RN 1017778-40-3

IUPAC Name: 1-(3-ethoxy-2,4-difluorophenyl)ethan-1-one
Synonyms: 1-(3-ethoxy-2,4-difluorophenyl)ethanone
Molecular Weight (g/mol): 200.19
Molecular Formula: C10H10F2O2
InChi Key: WKSSJUBUQZDDLP-UHFFFAOYSA-N
SMILES: CCOC1=C(F)C=CC(C(C)=O)=C1F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results