CAS RN 1017777-72-8
IUPAC Name:
6-chloro-2-fluoro-3-methoxybenzonitrile
Synonyms:
6-chloro-2-fluoro-3-methoxybenzonitrile
Molecular Weight (g/mol):
185.58
Molecular Formula:
C8H5ClFNO
InChi Key:
MRGPSVJVQWPCNB-UHFFFAOYSA-N
SMILES:
COC1=C(F)C(C#N)=C(Cl)C=C1
1
–
1
of
1
results
1
6-Chloro-2-fluoro-3-methoxybenzonitrile, 97%, Thermo Scientific™
CAS: 1017777-72-8 Molecular Formula: C8H5ClFNO Molecular Weight (g/mol): 185.58 MDL Number: MFCD09832269 InChI Key: MRGPSVJVQWPCNB-UHFFFAOYSA-N Synonym: 6-chloro-2-fluoro-3-methoxybenzenecarbonitrile,6-chloro-2-fluoro-m-anisonitrile PubChem CID: 46737506 IUPAC Name: 6-chloro-2-fluoro-3-methoxybenzonitrile SMILES: COC1=C(F)C(C#N)=C(Cl)C=C1