accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1016748-73-4

CH3H3CNOCl

CAS RN 1016748-73-4

IUPAC Name: N-benzyl-4-chloro-N-(propan-2-yl)butanamide
Synonyms: N-benzyl-4-chloro-N-isopropylbutanamide
Molecular Weight (g/mol): 253.77
Molecular Formula: C14H20ClNO
InChi Key: BEGDLNHPGOYQHE-UHFFFAOYSA-N
SMILES: CC(C)N(CC1=CC=CC=C1)C(=O)CCCCl

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results