accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1011-15-0

IUPAC Name: 1-(2-fluorophenyl)piperazine
Synonyms: 1-(2-fluorophenyl)piperazine
Molecular Weight (g/mol): 180.23
Molecular Formula: C10H13FN2
InChi Key: IVTZRJKKXSKXKO-UHFFFAOYSA-N
SMILES: FC1=CC=CC=C1N1CCNCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results