accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 100587-94-8

NOOONOOONOOOLa

CAS RN 100587-94-8

IUPAC Name: lanthanum(3+) hydrate trinitrate
Synonyms: lanthanum(3+) hydrate trinitrate
Molecular Weight (g/mol): 342.93
Molecular Formula: H2LaN3O10
InChi Key: HVMFKXBHFRRAAD-UHFFFAOYSA-N
SMILES: O.[La+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results