accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

H3COOHOFFF

CAS RN 1005205-20-8

IUPAC Name: 3-[4-methoxy-3-(trifluoromethyl)phenyl]propanoic acid
Synonyms: 3-[4-methoxy-3-(trifluoromethyl)phenyl]propanoic acid
Molecular Weight (g/mol): 248.2
Molecular Formula: C11H11F3O3
InChi Key: BZGRRQPEHHXJNW-UHFFFAOYSA-N
SMILES: COC1=C(C=C(CCC(O)=O)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results