accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CH3NFF

CAS RN 1003708-74-4

IUPAC Name: 3,5-difluoro-2-methylbenzonitrile
Synonyms: 3,5-difluoro-2-methylbenzonitrile
Molecular Weight (g/mol): 153.13
Molecular Formula: C8H5F2N
InChi Key: JHMDKULXRYFRBP-UHFFFAOYSA-N
SMILES: CC1=C(F)C=C(F)C=C1C#N
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results