accessibility menu, dialog, popup

UserName

New Feature

You can now browse the site in English or Deutsch.

CAS RN 1003706-87-3

NNBrF

CAS RN 1003706-87-3

IUPAC Name: 4-bromo-5-fluoropyrimidine
Synonyms: 4-bromo-5-fluoropyrimidine
Molecular Weight (g/mol): 176.98
Molecular Formula: C4H2BrFN2
InChi Key: PPJOFMHOVLXHPA-UHFFFAOYSA-N
SMILES: FC1=CN=CN=C1Br

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results