Every Space On the Bench - Frequently Asked Questions
When selecting a benchtop centrifuge, consider factors such as capacity, speed range, rotor compatibility, and safety features. Assess your lab's specific needs to find the best fit.
Microcentrifuges are specialized centrifuges designed for small volume samples, typically ranging from microliters to milliliters. They operate at high speeds to generate centrifugal force for applications such as DNA/RNA isolation, protein precipitation, and cell pelleting, making them ideal for molecular biology and biochemistry research.
When selecting a microcentrifuge, consider factors such as maximum speed, rotor compatibility, sample capacity, and safety features. Assessing your lab's specific needs and applications will help you choose the right model to ensure efficient and reliable sample processing.
No, a laboratory freezer is not the same as a normal household freezer. Lab freezers are specifically designed for storing temperature-sensitive materials and samples used in scientific research, healthcare, or industrial applications. They often have more precise temperature control, wider temperature ranges, and advanced features like alarms, data logging, and remote monitoring. Lab freezers may also have specialized designs, such as explosion-proof or flammable storage capabilities, to ensure safety in laboratory settings. In contrast, normal freezers are typically used for general food storage and may not provide the same level of temperature control and features required for scientific purposes.
Vortex mixers utilize rapid orbital shaking to mix small volumes of liquids in laboratory vessels such as tubes, vials, and microplates. They are commonly used for various applications, including sample re-suspension, cell culture mixing, PCR preparation, and enzyme reactions, providing quick and efficient mixing without the need for manual agitation.
When selecting a vortex mixer, consider features such as variable speed control, interchangeable attachments for different tube sizes, timer functions, and stability during operation. Additionally, assess factors like noise level, footprint, and durability to ensure compatibility with your lab's workflow and space constraints.
Benchtop pH meters are stationary instruments designed for precise and accurate measurement of pH in laboratory settings. They typically offer advanced features such as temperature compensation, data logging, and compatibility with specialized electrodes, making them suitable for demanding applications requiring high accuracy and reproducibility.
Regular calibration using standard buffer solutions is essential to maintain the accuracy and reliability of benchtop pH meters. Follow manufacturer guidelines for calibration procedures and frequency. Additionally, proper maintenance, including cleaning electrodes and storing the meter correctly, will prolong its lifespan and ensure consistent results.