Volumetric Flasks Banner
Volumetric Flasks
Volumetric flasks are essential laboratory vessels designed for the accurate preparation, dilution and measurement of solutions. Available in a wide range of capacities, materials and calibration classes, volumetric flasks help ensure reproducible analytical results across research, pharmaceutical, environmental, industrial and educational laboratories. This category includes high-quality options from Fisherbrand™, DWK™, Corning™, Pyrex™ and Thermo Scientific™ brands, offering reliable performance for routine and regulated laboratory workflows.
X5 100mL Fisherbrand™ Clear Borosilicate Glass Class A Volumetric Flask with Stopper
Manufactured from Borosilicate Glass and complies with ISO 1042 and DIN 12664.
Fisherbrand™ Clear Borosilicate Glass Class A Volumetric Flask with Stopper
X5 10mL Fisherbrand™ Clear Borosilicate Glass Class A Volumetric Flask with Stopper
Manufactured from Borosilicate Glass and complies with ISO 1042 and DIN 12664. 
Fisherbrand™ Amber Borosilicate Glass Class A Volumetric Flask with Stopper
X5 50mL Fisherbrand™ Clear Borosilicate Glass Class A Volumetric Flask with Stopper
Manufactured from Borosilicate Glass and complies with ISO 1042 and DIN 12664.
BRAND™ Blaubrand™ Class A Borosilicate Glass Volumetric Flasks with Glass Stopper
Save Now - Exclusive Deals
FAQ
A volumetric flask is used to prepare solutions to a precise volume. Laboratories use volumetric flasks for analytical chemistry, calibration procedures and solution preparation where accuracy and repeatability are essential.
Class A volumetric flasks provide tighter manufacturing tolerances and greater measurement accuracy than Class B flasks. They are commonly selected for regulated, analytical and quality-control applications.
Borosilicate glass is widely preferred because it offers excellent chemical resistance, thermal stability and durability. Plastic volumetric flasks may be selected when break resistance is a priority.
Volumetric flasks are used in research laboratories, pharmaceutical facilities, environmental testing laboratories, academic institutions and industrial quality-control environments.
These manufacturers offer high-quality volumetric flasks designed for reliable performance, certified accuracy and long-term durability, helping laboratories maintain consistent analytical results.
Top Brands