Chromatography Autosampler Vial Caps
Chromatography autosampler vial caps are essential for maintaining sample integrity throughout analytical workflows. Designed for HPLC, UHPLC and GC applications, these caps provide reliable sealing performance, minimize contamination risks and support reproducible analytical results. The Fisher Scientific channel offers a comprehensive range of chromatography vial cap solutions from the Thermo Scientific, Fisherbrand and DWK brands/portfolios, including screw caps, snap caps, crimp caps and septa configurations suitable for routine and advanced laboratory environments.
Fisherbrand™ 9mm PP Short Thread Seal, Blue, Center Hole, Assembled Septum
Blue, Red, White, Silicone/PTFE, 1 mm, Pack of 1000.
Thermo Scientific™ SureSTART™ 9 mm Screw Caps, Level 2 High-Throughput Applications
Blue, White Silicone/Red PTFE, 100 Pk., Pack of 100.
Fisherbrand™ 24mm PP Screw Seal, White, Closed Top, 24-400 Thread, Assembled Septum
White, Gray, Red, PTFE/Butyl, 2.5 mm, Pack of 100.
Save Now - Exclusive Deals
Must Have
FAQ
Chromatography autosampler vial caps seal sample vials used in HPLC, UHPLC and GC workflows. They help prevent evaporation, contamination and sample loss while supporting consistent analytical performance.
Select a cap based on vial size, closure type, septa material and instrument compatibility. The Thermo Scientific, Fisherbrand and DWK brands/portfolios offer options for routine and advanced chromatography methods.
Silicone/PTFE septa are widely used because they provide chemical resistance, low extractables and reliable resealing performance during repeated injections.
Screw caps are commonly preferred for routine laboratory workflows because they are easy to apply, convenient to remove and provide dependable sealing performance.
High-quality vial caps help maintain sample integrity, reduce contamination risks and support reproducible chromatographic results, contributing to more reliable analytical data.
Top Brands