Learn More
Vilber Lourmat™ UV Tube, 15w
Catalog No. 11872475
Ansicht ändern
Click to view available options
Wavelength:
254 nm
Packungsgröße:
1 Stück
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Wavelength Packungsgröße
11872475 254 nm 1 Stück
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11872475 Lieferant Vilber Lourmat™ Lieferanten-Nr. 907521511
  • ONLINE EXCLUSIVE PRICE
    78.80 €
    65.14 € / Pack of 1
    Save 13.66 €
    17% Off
Estimated Shipment: 07-01-2027