missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate concentrate, For 1L standard solution, 0.1 M ZnSO4, Honeywell Fluka™

Catalog Number p-7122057
Click to view available options
Quantity:
1 Ea.
6 x 1 Ea.
7733-02-0
O4SZn
161.44
MFCD00011302
NWONKYPBYAMBJT-UHFFFAOYSA-L
zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
24424
CHEBI:35176
zinc;sulfate
[Zn++].[O-]S([O-])(=O)=O
7733-02-0
O4SZn
161.44
MFCD00011302
NWONKYPBYAMBJT-UHFFFAOYSA-L
zinc sulfate, zinc sulphate, zincate, white vitriol, zinc sulfate 1:1, complexonat, bonazen, medizinc, optised, optraex
24424
CHEBI:35176
zinc;sulfate
[Zn++].[O-]S([O-])(=O)=O

for 1L standard solution, 0.1 M ZnSO4

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 1 Ea.
Linear Formula ZnSO4
UN Number 3082
Formula Weight 161.45g/mol
Packaging Ampoule
Chemical Name or Material Zinc Sulfate Concentrate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'