missing translation for 'onlineSavingsMsg'
Learn More

ZDHHC15 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA551975

Additional Details : Weight : 0.02000kg

Product Code. 15729930

  • 514.00 € / 100µL
Estimated Shipment: 11-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: VSRNKTTLEA FCTPVFTSGP EKNGFNLGFI KNIQQVFGDK KKFWLIPIGS SPGDGHSFPM RSMNESQNPL LANEETWEDN EDDNQ Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 95%.

ZDHHC15 Polyclonal antibody specifically detects ZDHHC15 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).
Specifications

Specifications

ZDHHC15
Polyclonal
Unconjugated
ZDHHC15
6030457O13Rik; DHHC-15; membrane-associated DHHC15 zinc finger protein; MRX91; Palmitoyltransferase ZDHHC15; UNQ1969/PRO4501; Zdhhc15; Zinc finger DHHC domain-containing protein 15; zinc finger DHHC-type containing 15; zinc finger, DHHC domain containing 15; zinc finger, DHHC-type containing 15
Rabbit
Antigen affinity chromatography
RUO
158866
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunocytochemistry, Immunohistochemistry (Paraffin)
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96MV8
ZDHHC15
Recombinant protein corresponding to Human ZDHHC15
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Research Use Only