missing translation for 'onlineSavingsMsg'
Learn More

Zatebradine hydrochloride, MedChemExpress

Product Code. p-200096218
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488367

Brand: Medchem Express HY13422

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zatebradine (UL-FS-49 (free base)) is a potent inhibitor of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels with an IC50 values 1.96 μM. Zatebradine blocks the slow inward current through human HCN1, HCN2, HCN3 and HCN4 channels, with IC50 values of 1.83 μM, 2.21 μM, 1.90 μM and 1.88 μM, respectively.

Zatebradine (UL-FS-49 (free base)) is a potent inhibitor of hyperpolarization-activated cyclic nucleotide-gated (HCN) channels with an IC50 values 1.96 μM. Zatebradine blocks the slow inward current through human HCN1, HCN2, HCN3 and HCN4 channels, with IC50 values of 1.83 μM, 2.21 μM, 1.90 μM and 1.88 μM, respectively.

Specifications

Chemical Name or Material Zatebradine hydrochloride
CAS 91940-87-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C26H37ClN2O5
Quantity 5 mg
Synonym UL-FS-49 UL-FS-49CL
Solubility Information DMSO : 100 mg/mL (202.82 mM; Need ultrasonic) ∣H2O : 11.11 mg/mL (22.53 mM; Need ultrasonic)
SMILES O=C1N(CCCN(CCC2=CC=C(OC)C(OC)=C2)C)CCC3=CC(OC)=C(OC)C=C3C1.Cl
Molecular Weight (g/mol) 493.04
Formula Weight 493.04
Percent Purity 98.03%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.