Learn More
Waters Corporation XBridge Prep C18 Column
As a workhorse for HPLC method development, XBridge BEH C18 sorbent can be used over an entire range of mobile phase pH (1 to 12).
772.50 € - 27900.00 €
Specifications
Max. Pressure | 6000 psi (415 Bar) |
---|---|
Particle Shape | Spherical |
Pore Size | 130 Å |
Quantity | 1/Ea. |
USP Type | L1 |
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Length (Metric) | Particle Size | Price | Quantity & Availability | |||
13212930
|
Waters Corporation
186002979 |
19 mm | 150 mm | 5 μm |
6030.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13202930
|
Waters Corporation
186002978 |
19 mm | 100 mm | 5 μm |
5040.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
10004293
|
Waters Corporation
186002980 |
30 mm | 50 mm | 5 μm |
6270.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13222930
|
Waters Corporation
186002981 |
30 mm | 75 mm | 5 μm |
7995.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
10470495
|
Waters Corporation
186002982 |
30 mm | 100 mm | 5 μm |
9255.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13184999
|
Waters Corporation
186003284 |
30 mm | 150 mm | 5 μm |
11595.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
10165384
|
Waters Corporation
186003892 |
19 mm | 10 mm | 10 μm |
772.50 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
10729627
|
Waters Corporation
186004021 |
19 mm | 250 mm | 5 μm |
7740.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13273240
|
Waters Corporation
186003893 |
19 mm | 50 mm | 10 μm |
2925.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13283240
|
Waters Corporation
186003894 |
19 mm | 150 mm | 10 μm |
5760.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
10083834
|
Waters Corporation
186003895 |
19 mm | 250 mm | 10 μm |
7290.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13293240
|
Waters Corporation
186003896 |
30 mm | 150 mm | 10 μm |
13050.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13203250
|
Waters Corporation
186003897 |
30 mm | 250 mm | 10 μm |
20400.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13213250
|
Waters Corporation
186003898 |
50 mm | 50 mm | 10 μm |
11790.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13223250
|
Waters Corporation
186003899 |
50 mm | 150 mm | 10 μm |
21900.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13233250
|
Waters Corporation
186003900 |
50 mm | 250 mm | 10 μm |
27900.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13243250
|
Waters Corporation
186003901 |
19 mm | 100 mm | 10 μm |
4860.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13253250
|
Waters Corporation
186003902 |
50 mm | 100 mm | 10 μm |
16050.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13233260
|
Waters Corporation
186003930 |
30 mm | 100 mm | 10 μm |
9540.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13243260
|
Waters Corporation
186003933 |
50 mm | 50 mm | 5 μm |
11745.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13283260
|
Waters Corporation
186003937 |
50 mm | 100 mm | 5 μm |
16050.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
10678974
|
Waters Corporation
186002977 |
19 mm | 50 mm | 5 μm |
3015.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13263330
|
Waters Corporation
186004025 |
30 mm | 250 mm | 5 μm |
16350.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13283360
|
Waters Corporation
186004107 |
50 mm | 250 mm | 5 μm |
25500.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
13223260
|
Waters Corporation
186003929 |
50 mm | 150 mm | 5 μm |
20250.00 €
Each |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||
Description
- The trifunctionally bonded C18 ligand provides you with a universal column for most HPLC separations and gives you the widest usable pH range, superior low pH stability, and ultra-low column bleed.
Spécification
6000 psi (415 Bar) | |
130 Å | |
L1 | |
Column |
Spherical | |
1/Ea. | |
Yes |