missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ WHSC1 (Human) Recombinant Protein
Human WHSC1 full-length ORF ( NP_579889.1, 1 a.a. - 647 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00007468-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MEFSIKQSPLSVQSVVKCIKMKQAPEILGSANGKTPSCEVNRECSVFLSKAQLSSSLQEGVMQKFNGHDALPFIPADKLKDLTSRVFNGEPGAHDAKLRFESQEMKGIGTPPNTTPIKNGSPEIKLKITKTYMNGKPLFESSICGDSAADVSQSEENGQKPENKARRNRKRSIKYDSLLEQGLVEAALVSKISSPSDKKIPAKKESCPNTGRDKDHLLKYNVGDLVWSKVSGYPWWPCMVSADPLLHSYTKLKGQKKSARQYHVQFFGDAPERAWIFEKSLVAFEGEGQFEKLCQESAKQAPTKAEKIKLLKPISGKLRAQWEMGIVQAEEAASMSVEERKAKFTFLYVGDQLHLNPQVAKEAGIAAESLGEMAESSGVSEEAAENPKSVREECIPMKRRRRAKLCSSAETLESHPDIGKSTPQKTAEADPRRGVGSPPGRKKTTVSMPRSRKGDAASQFLVFCQKHRDEVVAEHPDASGEEIEELLRSQWSLLSEKQRARYNTKFALVAPVQAEEDSGNVNGKKRNHTKRIQDPTEDAEAEDTPRKRLRTDKHSLRKRDTITDKTARTSSYKAMEAASSLKSQAATKNLSDACKPLKKRNRASTAASSALGFSKSSSPSASLTENELLWEPTPVKLDLNPAALYCT
Specifications
NP_579889.1 | |
Wolf-Hirschhorn syndrome candidate 1 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
WHSC1 | |
GST |
7468 | |
Wheat germ expression system | |
10 μg | |
FLJ23286, KIAA1090, MGC176638, MMSET, NSD2, REIIBP, TRX5, WHS | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |