Learn More
DWK Life Sciences 520 Bottle-Top Dispenser Reservoirs
Large selection of glass and Polyethylene reagent bottles, each supplied with one screw cap. Perfectly adapted with Calibrex™ bottle top dispensers.
28.55 € - 235.00 €
Specifications
Product Type | Reagent Bottle |
---|---|
For Use With (Equipment) | 520 model bottletop dispenser |
Product Code | Brand | Volume (Metric) | Color | Diameter (Metric) Outer Neck | Shape | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Color | Diameter (Metric) Outer Neck | Shape | Price | Quantity & Availability | ||
12457191
|
DWK Life Sciences
844029 |
1000 mL | Amber | 45 mm | Square |
45.05 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
12467191
|
DWK Life Sciences
844030 |
2500 mL | Amber | 45 mm | Round |
75.00 €
Each |
Estimated Shipment: 01-08-2024
Log in to see stock. |
||
12427191
|
DWK Life Sciences
844025 |
250 mL | Amber | 32 mm | Square |
28.55 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
12437191
|
DWK Life Sciences
844026 |
500 mL | Clear | 32 mm | Round |
235.00 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
12447191
|
DWK Life Sciences
844027 |
500 mL | Amber | 32 mm | Square |
57.30 €
Each |
Estimated Shipment: 13-08-2024
Log in to see stock. |
||
Description
- For use with Wheaton 520 bottle-top dispensers
- Cat. No. 13-706-8, made of clear borosilicate glass; with graduations
- All others ungraduated and made of amber glass (13-706-9, Plastisol™-coated amber glass)
Specifications
Reagent Bottle | |
520 model bottletop dispenser |