Learn More
Saint Gobain Life Sciences Chemware™ PFA Narrow Mouth Wash Bottle
Product Code. 11744528
Click to view available options
Capacity (Metric):
100 mL
250 mL
500 mL
1000 mL
Contains the lowest concentration of trace metal ions of all fluoropolymers making it ideal for the ultra-pure requirements of environmental analytical work. Chemware™ PFA Narrow Mouth Wash Bottle can be used to dispense all types of liquids.
  • Molded-in graduation marks
  • Flexible nozzle with 360° swivel; small orifice tip allows accurate content dispensing
  • Inert to all chemicals
  • Leak-proof design
  • Transparent and break resistant
  • Non-stick, easy to clean and provide superior flex life and excellent mechanical strength
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 1000 mL
Capacity (English) 33.81 oz.
Diameter (Metric) Inner Neck 25.5 mm
Material PFA
Closure Included Yes
Graduated Yes
Mouth Narrow
Height (Metric) 315.1 mm
Product Type Wash Bottle
Height (English) 12.4 in.