Learn More
Macherey-Nagel™ Visocolor™ HE Titrationstestkit, Ersatzspritze
Ersetzen Sie die Titrationsspritze aus Testkits. Ersatzspritzen für Macherey-Nagel™ Visocolor™ HE Titrations-Testkits werden bei HE-Tests verwendet, um selbst die niedrigsten Grenzwerte zu bestimmen. Die Graduierungsmarkierungen zeigen den Gehalt des getesteten Parameters an, die der Menge des verwendeten Titrationsmittels entspricht.
Marke: Macherey-Nagel™ 915407
Weitere Details : Gewicht : 0.50000kg
Beschreibung
Visocolor™ HE test kits provide highly precise analysis via exact dosing of the reagents, as well as by compensating for turbidity and color. The sensitivity of Visocolor™ HE is 10 to 100 times higher compared to other Visocolor tests. Each test kit is economically priced and provides convenient handling.
- High sensitive volumetric analysis via visual evaluation
- Narrow syringe with gradation marks for easy, accurate readings right on the barrel
- Gradations show equivalent analyte level based on titrant required for color change
- Sharper color change due to separated dropping reagents
- Refill packs and replacement titration syringes available
- Single reagent bottles with clear dosing instructions
- Delivered in a robust box with plastic inlay and easy-to-understand instructions
Spezifikation
OH- | |
Alkalinity determination | |
Graduated Syringe | |
Titration syringe, dropping tips | |
1 Each | |
Colorimetric titration |
Clear barrel, black graduation marks, white plunger with black o-ring | |
Visocolor™ HE Test Kit for Alkalinity AL 7 (915 007) | |
0.2mmol/L gradation mark | |
200 | |
0.2 to 7.2mmol/L OH- | |
Titration syringe |