missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Vinblastine sulfate, 96.0-102.0%

Catalog Number p-3779872
Click to view available options
Quantity:
5 mg
25 mg
Packaging:
Glass bottle
This item is not returnable. View return policy
143-67-9
C46H60N4O13S
909.06
MFCD08706468,MFCD00082457,MFCD08706468
KDQAABAKXDWYSZ-JKDPCDLQSA-N
vinblastine sulfate
131709225
methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC
This item is not returnable. View return policy
143-67-9
C46H60N4O13S
909.06
MFCD08706468,MFCD00082457,MFCD08706468
KDQAABAKXDWYSZ-JKDPCDLQSA-N
vinblastine sulfate
131709225
methyl (1R,9R,10S,11R,12R,19R)-11-(acetyloxy)-12-ethyl-4-[(13S,15S,17S)-17-ethyl-17-hydroxy-13-(methoxycarbonyl)-1,11-diazatetracyclo[13.3.1.0⁴,¹².0⁵,¹⁰]nonadeca-4(12),5,7,9-tetraen-13-yl]-10-hydroxy-5-methoxy-8-methyl-8,16-diazapentacyclo[10.6.1.0¹,⁹.0²,⁷.0¹⁶,¹⁹]nonadeca-2(7),3,5,13-tetraene-10-carboxylate; sulfuric acid
OS(O)(=O)=O.CC[C@]1(O)C[C@H]2CN(C1)CCC1=C(NC3=CC=CC=C13)[C@@](C2)(C(=O)OC)C1=CC2=C(C=C1OC)N(C)[C@@H]1[C@]22CCN3CC=C[C@](CC)([C@@H]23)[C@@H](OC(C)=O)[C@]1(O)C(=O)OC

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 267.0°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96.0 to 102% (HPLC)
Packaging Glass bottle
Quantity 5 mg
Merck Index 15, 10179
Specific Rotation Condition −21.70° (21°C c=2,CH3OH)
Specific Rotation -21.7
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohols,acetone,ethyl acetate,,chloroform,practically insoluble in petroleum ether
Formula Weight 909.05
Percent Purity 96.0-102%
Physical Form Crystalline Powder
Chemical Name or Material Vinblastine sulfate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'