missing translation for 'onlineSavingsMsg'
Learn More

Saint Gobain Life Sciences Versilic™ Sealing Caps

Sterilizable Versilic™ Sealing Caps for test tubes.

Brand:  Saint Gobain Life Sciences 162014-100

 View more versions of this product

Product Code. 11744618

  • 168.00 € / Pack of 100
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For test tubes.
  • Easy to install.
  • Good chemical inertness.
  • Sterilizable.
  • Temperatures between -76°F and +428°F (-60°C and +220°C).
TRUSTED_SUSTAINABILITY
Specifications

Specifications

14 mm
Plastic
Non-sterile
Versilic Cap
100 Pk.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions