Learn More
Fisherbrand VCpro601, Vacuum Control Box, 1100 - 1 mbar
Brand:  Fisherbrand 600100
VCpro601, Vacuum Control Box, 1100 - 1 mbar
Product Code. 16675922
  • ONLINE EXCLUSIVE PRICE
    2385.00 €
    2267.00 € / Each
    Save 118.00 €
    4% Off
Only null left
Estimated Shipment: 09-01-2026