Learn More
Honeywell™ Uvex™ Astrospec 3000 UV
Click to view available options
Frame Color: Yellow and black
Description
Specifications
Certifications/Compliance | EN 170 |
Lens Type | Uvex™ supravision sapphire |
Lens Tint | Clear |
Frame Color | Yellow, Black |
Temple Color | Yellow |
Frame Type | Shield-lens spectacle |