missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Triton|r X-100, Electrophoresis Reagent

Catalog Number p-7057520
Click to view available options
Quantity:
100 mL
2500 mL
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
9002-93-1
C16H26O2
Octoxynol-9; Polyethylene glycol tert-octylphenyl ether
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.
9002-93-1
C16H26O2
Octoxynol-9; Polyethylene glycol tert-octylphenyl ether

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Colorless to Yellow
Content And Storage Ambient temperatures
Density 1.061 g/mL
Formula Weight ca. 625
MDL Number MFCD00132505
Quantity 100 mL
Melting Point ∼5°C
Odor Mild
InChI Key JYCQQPHGFMYQCF-UHFFFAOYSA-N
SMILES CC(C)(C)CC(C)(C)C1=CC=C(OCCO)C=C1
IUPAC Name 2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethan-1-ol
Molecular Weight (g/mol) 250.38
Form Liquid
Chemical Name or Material Triton™ X-100
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'