missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tritolyl phosphate, 99%, mixture of isomers, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
500 g
2.5 kg
Packaging:
Glass bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Melting Point | -30°C |
Color | White |
Density | 1.1600g/mL |
Boiling Point | 265°C (10.0 mmHg) |
Flash Point | 234°C |
Infrared Spectrum | Authentic |
Assay Percent Range | 98.5% min. (sum of isomers) (GC) |
Packaging | Glass bottle |
Linear Formula | (CH3C6H4O)3PO |
Refractive Index | 1.5550 to 1.557 |
Show More |
Safety and Handling
Product Identifier
- Tritolyl phosphate, mixture of isomers
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Reproductive toxicity Category 2
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H361fd-Suspected of damaging fertility. Suspected of damaging the unborn child.
- P201-Obtain special instructions before use.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P391-Collect spillage.
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'