missing translation for 'onlineSavingsMsg'
Learn More

Triphenylcarbenium hexachloroantimonate, 99%, Thermo Scientific™

Catalog Number p-7027955
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
1586-91-0
C19H15Cl6Sb
577.79
MFCD00013126
GASWLBDVMZXHOP-UHFFFAOYSA-H
tritylium hexachloroantimonate, triphenylcarbenium hexachloroantimonate, trityl hexachloroantimonate, hexachlorostibanuide; triphenylmethylium
16688166
Cl[Sb-](Cl)(Cl)(Cl)(Cl)Cl.C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1
This item is not returnable. View return policy
1586-91-0
C19H15Cl6Sb
577.79
MFCD00013126
GASWLBDVMZXHOP-UHFFFAOYSA-H
tritylium hexachloroantimonate, triphenylcarbenium hexachloroantimonate, trityl hexachloroantimonate, hexachlorostibanuide; triphenylmethylium
16688166
Cl[Sb-](Cl)(Cl)(Cl)(Cl)Cl.C1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY
Melting Point ∼220°C (decomposition)
Odor Odorless
Linear Formula (C6H5)3CSbCl6
Quantity 5 g
UN Number UN3261
Beilstein 3817547
Sensitivity Moisture sensitive
IUPAC Name diphenylmethylbenzene;hexachloroantimony(1-)
Formula Weight 577.8
Percent Purity 99%
Chemical Name or Material Triphenylcarbenium hexachloroantimonate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'