missing translation for 'onlineSavingsMsg'
Learn More

Trimethyl-p-tolylsilane, 95%, Thermo Scientific Chemicals

Catalog Number p-7023884
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
3728-43-6
C10H16Si
164.323
MFCD00014850
QGHURGPPCGMAMZ-UHFFFAOYSA-N
trimethyl-p-tolylsilane, p-tolyltrimethylsilane, trimethyl 4-methylphenyl silane, trimethyl-p-tolyl silane, trimethyl p-tolyl silane, 4-trimethylsilyl toluene, silane, p-tolyl-trimethyl-,, 4-methylphenyl trimethylsilane
298149
trimethyl-(4-methylphenyl)silane
CC1=CC=C(C=C1)[Si](C)(C)C
This item is not returnable. View return policy
3728-43-6
C10H16Si
164.323
MFCD00014850
QGHURGPPCGMAMZ-UHFFFAOYSA-N
trimethyl-p-tolylsilane, p-tolyltrimethylsilane, trimethyl 4-methylphenyl silane, trimethyl-p-tolyl silane, trimethyl p-tolyl silane, 4-trimethylsilyl toluene, silane, p-tolyl-trimethyl-,, 4-methylphenyl trimethylsilane
298149
trimethyl-(4-methylphenyl)silane
CC1=CC=C(C=C1)[Si](C)(C)C

It is employed in the production of trimethyl-(4-trimethylsilanyl-benzyl)-silane by reacting with chloro-trimethyl-silane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the production of trimethyl-(4-trimethylsilanyl-benzyl)-silane by reacting with chloro-trimethyl-silane.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Protect against electrostatic charges. Keep ignition sources away.
TRUSTED_SUSTAINABILITY
Boiling Point 191°C to 192°C
Quantity 5 g
UN Number UN1993
Flash Point 46°C (115°F)
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 164.33
Percent Purity 97%
Refractive Index 1.491
Density 0.865
Chemical Name or Material Trimethyl-p-tolylsilane

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'