missing translation for 'onlineSavingsMsg'
Learn More

TRIM58 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  Thermo Scientific PA554856

Additional Details : Weight : 0.02000kg

Product Code. 15782781

  • 534.00 € / 100µL
Estimated Shipment: 10-04-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Immunogen sequence: EAGEISFYNV TDGSYIYTFN QLFSGLLRPY FFICDATPLI LPPTTIAGSG NWASRDHLDP ASDVRDDHL Highest antigen sequence identity to the following orthologs: Mouse - 68%, Rat - 71%.

TRIM58 Polyclonal antibody specifically detects TRIM58 in Human samples. It is validated for Western Blot.
Specifications

Specifications

TRIM58
Polyclonal
Unconjugated
TRIM58
BIA2; E3 ubiquitin-protein ligase TRIM58; Gm16; Gm1783; Protein BIA2; RGD1566174; RING-type E3 ubiquitin transferase TRIM58; TRIM58; tripartite motif containing 58; tripartite motif-containing 58; tripartite motif-containing protein 58
Rabbit
Antigen affinity chromatography
RUO
25893
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot
0.15 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q8NG06
TRIM58
Recombinant protein corresponding to Human TRIM58
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Research Use Only