missing translation for 'onlineSavingsMsg'
Learn More

Triethanolamine, 97%, Thermo Scientific Chemicals

Catalog Number p-203763
Click to view available options
Quantity:
100 mL
1 L
2.5 L
25 L
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
102-71-6
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
daltogen, triethanolamine, 2,2',2-nitrilotriethanol, thiofaco t-35, sterolamide, trihydroxytriethylamine, nitrilotriethanol, trolamine, tris 2-hydroxyethyl amine, triethylolamine
7618
CHEBI:28621
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
102-71-6
149.19
MFCD00002855
GSEJCLTVZPLZKY-UHFFFAOYSA-N
daltogen, triethanolamine, 2,2',2-nitrilotriethanol, thiofaco t-35, sterolamide, trihydroxytriethylamine, nitrilotriethanol, trolamine, tris 2-hydroxyethyl amine, triethylolamine
7618
CHEBI:28621

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Decomposition Information >325°C
Melting Point 21.0°C
Density 1.1245 g/mL
Boiling Point 360.0°C
Flash Point 190°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C6H15NO3
Linear Formula (HOCH2CH2)3N
Refractive Index 1.4840 to 1.4860
Quantity 100 mL
Fieser 01,1196
Merck Index 15, 9830
Specific Gravity 1.1245
Solubility Information Solubility in water: freely soluble. Other solubilities: miscible with alcohol,methanol and acetone,soluble in chloroform,slichtly soluble in benzene and ether,soluble 4.2% in benzene; 1.6% in ether; 0.4% in,carbon tetrachloride and < 0.1% in n-he
SMILES OCCN(CCO)CCO
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethan-1-ol
Viscosity 600 mPa.s (25°C)
Formula Weight 149.19
Vapor Pressure <0.01 mmHg (at 20°C)
Percent Purity 97%
Physical Form Oily Liquid
Chemical Name or Material Triethanolamine, 97%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'