missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Trichloroethylene, Honeywell
Click to view available options
Quantity:
1 L
2.5 L
Specifications
Density | 1.463g/mL at 25°C |
Ignition Point | 770°F |
Linear Formula | ClCH=CCl2 |
Quantity | 1 L |
UN Number | 1710 |
Beilstein | 1736782 |
Vapor Density | 4.5 (vs air) |
Formula Weight | 131.39g/mol |
Vapor Pressure | 61mmHg (20°C) |
Percent Purity | ≥99% |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'