missing translation for 'onlineSavingsMsg'
Learn More

trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid, 97%, Thermo Scientific Chemicals

Catalog Number 15427666
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
61935-48-6
C15H19NO4
277.32
MFCD03844616
RPJMLWMATNCSIS-STQMWFEESA-N
z-1,2-trans-achc-oh, trans-1-benzyloxycarbonyl-amino-cyclohexyl-2-carboxylic acid, 1s,2s-2-benzyloxy carbonyl amino cyclohexane-1-carboxylic acid
51341834
(1S,2S)-2-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
C1CCC(C(C1)C(=O)O)NC(=O)OCC2=CC=CC=C2
This item is not returnable. View return policy
61935-48-6
C15H19NO4
277.32
MFCD03844616
RPJMLWMATNCSIS-STQMWFEESA-N
z-1,2-trans-achc-oh, trans-1-benzyloxycarbonyl-amino-cyclohexyl-2-carboxylic acid, 1s,2s-2-benzyloxy carbonyl amino cyclohexane-1-carboxylic acid
51341834
(1S,2S)-2-(phenylmethoxycarbonylamino)cyclohexane-1-carboxylic acid
C1CCC(C(C1)C(=O)O)NC(=O)OCC2=CC=CC=C2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Formula Weight 277.31
Percent Purity 97%
Quantity 250 mg
Physical Form Solid
Chemical Name or Material trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'